主要文件
27159
Supelco
预组装螺纹口样品瓶,带酚醛树脂顶部开口瓶盖
volume 15 mL, clear glass vial, O.D. × H 21 mm × 70 mm, tan PTFE/silicone septum
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41122001
材料
black phenolic cap (open-top)
clear glass vial
tan PTFE/silicone septum
特點
closure type black phenolic hole cap
closure type screw top vial
包裝
pkg of 100 ea
外徑 × 高度
21 mm × 70 mm
容積
15 mL
接頭
thread for 18-400
正在寻找类似产品? 访问 产品对比指南
历史批次信息供参考:
分析证书(COA)
访问文档库
Anna Banel et al.
Analytical and bioanalytical chemistry, 399(9), 3299-3303 (2010-11-26)
In this study, a procedure was developed to determine short-chain alkane monocarboxylic acids (SCMAs) in aqueous samples using headspace solid-phase microextraction (HS-SPME) followed by gas chromatography (GC) coupled with mass spectrometry (MS). A Stabilwax-DA capillary column (30 m × 0.32-mm
Determination of glyphosate and (aminomethyl) phosphonic acid in soil, plant and animal matrixes, and water by capillary gas chromatography with mass-selective detection.
Alferness P and Iwata Y.
Journal of Agricultural and Food Chemistry, 42 (12), 2751-2759 (1994)
Stable hydrogen-isotope analysis of methyl chloride emitted from heated halophytic plants.
Markus G
Atmospheric Environment, 62, 584-592 (2012)