Z641987
1 of 10
CLS430829
Corning®50 mL离心管
CLS430828
Z728039
Hettich® swing-out rotor, 4 place
CLS430921
Corning ® 50 mL PP 离心管,自立式
Sigma-Aldrich
Z371092
透析袋夹
T2318
Greiner 离心管
CLS430290
CLS430291
916765
Tapered dispensing tips
GF29491061
聚四氟乙烯
Holds 8.5 mL tubes
pack of 8 ea
Hettich 5248
centrifugation: suitable
for use with 5051 (bucket )
正在寻找类似产品? 访问 产品对比指南
实验室设备