主要文件
Z329371
BRAND® magnetic stirring bar, PTFE, triangular
L 50 mm, bar diam. 12 mm
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41103806
NACRES:
NB.72
材料
triangular PTFE-coated
包裝
pack of 10
製造商/商標名
BRAND 137820
長度
50 mm
桿直徑
12 mm
正在寻找类似产品? 访问 产品对比指南
一般說明
Wedge shaped. Ideal for dissolving solids and mixing of sediments due to scraping action. Angular surface creates high turbulence even at low speeds.
其他說明
BR137820 (India)
法律資訊
BRAND is a registered trademark of BRAND GMBH + CO KG