主要文件
GE80648546
GE Ettan DALTsix gel caster
volume 1000 mL, pkg of 1-6 gels
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41105300
NACRES:
NB.22
包裝
pack of 1 ea
pkg of 1-6 gels
製造商/商標名
Cytiva 80-6485-46
Cytiva Article No. 80-6485-46
容積
1000 mL
一般說明
DALTsix Gel Caster holds up to six 1.0-mm or 1.5-mm cassettes with separator sheets for casting homogeneous or gradient gels. If fewer gels are to be cast, the unused volume is filled with blank cassette inserts and filler sheets.
法律資訊
Ettan is a trademark of Cytiva
历史批次信息供参考:
分析证书(COA)