主要文件
GE18-1153-01
Tricorn Bottom Unit
Tricorn 5 Column
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41115709
NACRES:
NB.43
特點
Tricorn 5 Column
製造商/商標名
Cytiva 18-1153-01
18-1153-10
正在寻找类似产品? 访问 产品对比指南
一般說明
Tricorn columns incorporate modern design and high performance to increase the user friendliness and reliability of the chromatography column. Tricorn columns are designed for high resolution liquid chromatography of biomolecules performed at medium to high pressures.
  • High performance glass columns designed for reliable high resolution, reproducibility and durability
  • New longer adaptor for increased adjustment of bed height
  • Modern, integrated design – simple to assemble, pack, operate, and maintain
  • Broad range of column dimensions – suitable for many different purification needs
  • Safety first – glass tubes coated with plastic film to protect laboratory personnel if the column is dropped or operated outside its specifications
  • Locking ring – prevents accidental compression of media bed
  • Separate packing connector – allows simple column packing for a broad range of media
历史批次信息供参考:
分析证书(COA)