主要文件
EP5920753002
Eppendorf® Adapter
for 50 mL Oak Ridge tubes for rotor S-4xUniversal- Large
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41103911
NACRES:
NB.82
特點
Holds 50 mL tubes
for 50 mL Oak Ridge tubes for rotor S-4xUniversal- Large
包裝
set of 2 pieces
製造商/商標名
Eppendorf® 5920753002
技術
centrifugation: suitable
相容性
for use with Universal-Large bucket
法律資訊
Eppendorf is a registered trademark of Eppendorf AG
历史批次信息供参考:
分析证书(COA)