主要文件
EP5895117007
Rotor for Eppendorf® 5920R Centrifuges
swing-bucket rotor, S-4x1000, aerosol tight, with 4 plate/tubes buckets, to hold, 4 x 750 mL plates/tubes, with Eppendorf QuickLock® lid
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41103909
NACRES:
NB.82
特點
to hold 4 x 750 mL plates/tubes
aerosol tight
swing-bucket rotor
with 4 plate/tubes buckets
with Eppendorf QuickLock® lid
包裝
pack of 1 ea
製造商/商標名
Eppendorf® 5895117007 (Global)
參數
3,428 × g max. RCF
技術
centrifugation: suitable
相容性
for use with centrifuge models 5920R
正在寻找类似产品? 访问 产品对比指南
一般說明
The large Centrifuge 5920 R is a benchtop centrifuge that combines exceptional capacity with a compact and ergonomic design. It has similar dimensions to refrigerated competitor models in the 3-liter segment, but has an outstanding capacity of up to 4 x 1,000 mL or 52 x 50 mL tubes. Optimize high-throughput centrifugation and make your daily work easier.
法律資訊
Eppendorf is a registered trademark of Eppendorf AG
Eppendorf QuickLock is a registered trademark of Eppendorf AG
历史批次信息供参考:
分析证书(COA)