Key Documents
EP5427710000
Eppendorf® Rotor FA-45-24-11-HS
incl. rotor lid, aerosol-tight, coated
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41103909
NACRES:
NB.82
特點
to hold 24 x 1.5/2.0 mL tubes
incl. rotor lid, aerosol-tight, coated
包裝
pack of 1 ea
製造商/商標名
Eppendorf® 5427710000
參數
175,000 max. RCF (30,130 xg)
技術
centrifugation: suitable
相容性
for use with centrifuge models 5430/5430R
法律資訊
Eppendorf is a registered trademark of Eppendorf AG