主要文件
BAF187453011
No-Wire Multi-Tube Racks
number of holes 35
, Holds 4 of 50 mL, 6 of 15 mL, and 25 of 1.5 mL Centrifuge Tubes
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41122804
NACRES:
NB.22
材料
white polypropylene
特點
autoclavable
Holds 25 x 1.5 mL tubes
Holds 4 x 50 mL tubes
Holds 6 x 15 ml tubes
包裝
pack of 1 ea
製造商/商標名
Bel-Art F18745-3011
正在寻找类似产品? 访问 产品对比指南
相关类别
一般說明
Each No-Wire Racks holds two or three different sizes of round or conical bottom centrifuge tubes. One rack holds four 50ml, six 15ml and twenty-five 1.5ml tubes (hole sizes, 30.2mm, 17.3mm and 11.4mm respectively), other rack holds six 50ml and twelve 15ml centrifuge tubes (holes size, 30.2mm and 17.3mm respectively). Easy to assemble. Dimensions are 246mm x 104mm x 64mm high (9.7" x 4.1" x 2.5"). The white polypropylene racks are submersible and steam autoclavable to 121ºC (250º F).
Store different size tubes of required reagents using one space saving rack
  • Ships flat - Easy to assemble
法律資訊
No-Wire is a trademark of SP Scienceware, Division of SP
历史批次信息供参考:
分析证书(COA)