AXYHGB153515
别名:
gel system accessory
1 of 10
AXYHGB1530MC15
Corning® Axygen® 30 well MC comb for use with 15 cm gel system
Sigma-Aldrich
C4800
Combs for standard vertical gels, 16.5 cm wide
219392
组织蛋白酶B底物III,荧光法
EP5308000003
Eppendorf® ThermoTop With Condens.Protect® Technology
Z274909
Kimax® weighing bottle, Style I
721328
碘乙酰胺-13C2,2-d2
G5507
半乳糖基转移酶 来源于牛奶
8.55057
Universal NovaTag® resin
G7396
β-葡萄糖醛酸酶 来源于大肠杆菌
257370
7-氨基-4-甲基香豆素
Corning HGB15-35-15
(1.5 mm)
Corning®
实验室容器与储存
实验室耗材合作伙伴
样品处理
历史批次信息供参考:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our 文件 section.