P35015
1 of 10
Millipore
P30799
T-line Cartridge Housing Bowl to Base Sanitary Clamp 3 in.
Supelco
PX1235
Piperidine
YY1412212-M
Cartridge Housing Clamp Tri-Clover 4 in.
HP01PPVT0
Optiseal Sanitary Cartridge Housing T-line
17403X1
Cartridge Housing Silicone Vent/Drain Valve Seal Kit
SAFC
C5467
EX-CELL ® ACF CHO 培养基
SPPS00004
In-line Cartridge Housing Bowl to Base Silicone O-ring
Sigma-Aldrich
CAS9BAC1P
用于大肠杆菌的Cas9 Lambda Red同源重组质粒
HP01PPVN0
Optiseal Sanitary Cartridge Housing In-line
S2563
Sphingolipid Ceramide N-Deacylase from Pseudomonas sp.
4 in.
for use with Series 2000 Sanitary Cartridge Housings
滤筒_无菌过滤_生物负荷控制-默克生命科学