HGTC150EP
1 of 10
Millipore
YY2004045
夹具1 1/2 in. TC
GBL726024
Grace Bio-Labs HybriSlip™ hybridization cover
MP0DPIL0TX
中试规模夹具的扩大套件,用于Pod滤器
MP0DPIL0T
中试规模夹具
Sigma-Aldrich
E2645
Epidermal Growth Factor Receptor human
SAFC
1.08643
Triton™ X-100
SAB4502616
Anti-NF-κB p65 (Acetyl-Lys310) antibody produced in rabbit
126654
Albumin, Human Serum, Non-denatured
8.55049
H-Thr-Gly-NovaSyn® TG resin
MIXPDRH0IST
混合干粉袋吊车
1.5 in.
for use with Series 2000 Sanitary Cartridge Housings