材料
borosilicate glass bottle
polypropylene cap
round bottle
接合處
Joints threaded neck (GL45)
無菌
non-sterile
特點
graduations
with cap
包裝
pkg of 10 ea
製造商/商標名
Sigma-Aldrich
容量
250 mL
正在寻找类似产品? 访问 产品对比指南
一般說明
Borosilicate glass, wide mouth, media/reagent bottles with polypropylene GL 45 thread screw cap with pour ring. Large marking spot and reference graduations.
法律資訊
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Global Trade Item Number
货号 | GTIN |
---|---|
Z741012-10EA | 4061833641859 |