主要文件
Z682365
Ace rotary evaporator condenser complete "V" assembly
plastic-coated glass, fits Büchi® models R-200/R-205 and R-114/R-144, includes 1 L receiving flask and clamp
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121800
NACRES:
VB.02
材料
plastic-coated glass
製造商/商標名
Büchi equivalent, 40602
正在寻找类似产品? 访问 产品对比指南
一般說明
Includes 1 L receiving flask and clamp.
The main glass condenser body with inner coil for vertical installation, O-ring, 1 L receiving flask, stainless steel clamp, GL threaded caps and hose barbs.
法律資訊
Büchi is a registered trademark of Büchi Labortechnik AG
历史批次信息供参考:
分析证书(COA)