主要文件
Z678775
FORTUNA® Polyfix® bottle top dispenser with amber glass cylinder
dispensing volume 10-50 mL
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121500
NACRES:
NB.01
製造商/商標名
Fortuna Graf 101.072-48
dispensing volume measuring range
10-50 mL
容積
10-50 mL
接頭
GL 40
GL 45
正在寻找类似产品? 访问 产品对比指南
一般說明
For dispensing light sensitive liquids, weak acids and alkalis
Features:
  • With amber dispensing cylinder made of glass, coated with PP-protection against breakage
  • Glass piston
  • Ergonomic finger protection cap
  • Fast and easy volume setting with additional fine adjustment
  • FIX adapter made of PP
  • Valve block made of PP, no metal springs in the valve - no corrosion
Supplied as FORTUNA Polyfix dispenser, 2 PP adapters GL40 + GL45 & 1 PTFE intake tube
法律資訊
FORTUNA is a registered trademark of Poulten & Graf Ltd.
POLYFIX is a registered trademark of Poulten & Graf Ltd.
历史批次信息供参考:
分析证书(COA)