製造商/商標名
Bel-Art F18737-0000
長度 × 寬度 × 高度
5 3/4 in. × 4 13/16 × 4 1/4
一般說明
Polypropylene test tube basket; 5 x 4 x 4 in, without lid. Ideal for rinsing, transport and storage of glassware. .Steam autoclavable at 121ºC (250ºF)
法律資訊
Lab-Aire is a registered trademark of SP Scienceware, a division of SP
其他客户在看
Global Trade Item Number
货号 | GTIN |
---|---|
Z673609-1EA | 4061832266367 |