製造商/商標名
Q Labtech
容積
4 oz , catch bottle
正在寻找类似产品? 访问 产品对比指南
一般說明
QianCap is a safety cap with an automatic pressure release and reseal mechanism which can prevent pressure bottles and tubes with Ace-Threds #15 and #25 from explosion due to excessive internal pressure. These systems contain all items needed for immediate use on pressure tubes and bottles with internal threads. QianCap is available in these three configurations:
1. QianCap safety cap
2. QianCap safety cap with 0-300 psig pressure gauge
3. QianCap safety cap gas purging set, with 0-300 psig pressure gauge
Components in all configurations
1. QianCap safety cap
2. QianCap safety cap with 0-300 psig pressure gauge
3. QianCap safety cap gas purging set, with 0-300 psig pressure gauge
Components in all configurations
- One PTFE Q-adapter
- One Simriz® O-ring
- Ten septa - 12 mL
- One pressure adapter-60
- One cap
- One needle adapter
- One deflected point needle
- One catch bottle
法律資訊
QianCap is a trademark of Q Labtech LLC
Simriz is a registered trademark of Carl Freudenburg KG
其他客户在看
Global Trade Item Number
货号 | GTIN |
---|---|
Z568724-1EA | 4061833496169 |