製造商/商標名
Bel-Art F368290001
長度
46 1/4 in.
一般說明
Reaches deep inside glass vessels without scratching. The rigid nylon scraper blade (7 in. L × 4 1/2 in. W) is molded to a tubular, fiberglass shaft having a ribbed hand grip.
法律資訊
Scienceware is a registered trademark of SP Scienceware, a division of SP
其他客户在看
Global Trade Item Number
货号 | GTIN |
---|---|
Z225142-1EA | 4061832257730 |