材料
stainless steel
接合處
joint SJ35/25
無菌
non-sterile
包裝
pack of 1
一般說明
Use with flash-chromatography ball jointed assemblies and rotary evaporator receiver flasks.
其他客户在看
Global Trade Item Number
货号 | GTIN |
---|---|
Z224871-1EA | 4061833304167 |