Z220302
1 of 10
Sigma-Aldrich
Z166898
Aldrich® 气敏化学玻璃实验室套件
Z173088
Solvent storage flask with glass stopcock on side-arm
Z562491
LabJaws™ regular clamp holder
Z107174
Lattice-rod clamp
Millipore
XX1014705
Millipore真空过滤侧臂烧瓶
Z122548
Aldrich® student glass lab kit
Z173495
Addition funnel with Rotaflo® stopcock, and glass stopcock on side-arm
Z173096
XX1514706
Z243620
Aldrich clamp holder
pear-shaped single-neck stainless steel
Q
male joint: ST/NS14/20
non-sterile
closure type Standard Taper Joint
pack of 1
100 mL
正在寻找类似产品? 访问 产品对比指南
样品处理
历史批次信息供参考:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our 文件 section.
如需帮助,请联系 客户支持
在文件库中查找您最近购买产品的文档。
访问文档库
1 of 4
Z173126
Reaction and storage tube with glass stopcock on side-arm
Z516015
Schlenk reaction and storage tube
Z220434
Sealing tube
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.