主要文件
Z189928
Sigma-Aldrich
Mercury collector replacement pads
pkg of 12 ea
登录查看公司和协议定价
About This Item
分類程式碼代碼:
47131905
NACRES:
NB.35
包裝
pkg of 12 ea
製造商/商標名
Bel-Art
一般說明
12 Replacement pads for the Mercury Collector spill kit. Uset o collect spilled mercury quickly, just unscrew the lid with the foam pad, and press pad firmly onto the spill. This pressure forces the mercury into the pad. When the lid is screwed back onto the jar, the pad is compressed against the jar’s freestanding perforated plate, releasing the mercury into the jar bottom.
特點和優勢
  • Use recommended disposal method
  • Replacement pads should be affixed with rubber cement
历史批次信息供参考:
分析证书(COA)