主要文件
Z108359
Leather gloves
size XL
登录查看公司和协议定价
About This Item
分類程式碼代碼:
46181504
NACRES:
NB.35
材料
leather
尺寸
XL
無菌
non-sterile
特點
latex free
powder-free
reusable
包裝
pack of 1 pair
製造商/商標名
Sigma-Aldrich
正在寻找类似产品? 访问 产品对比指南
相关类别
一般說明
Constructed of specially treated leather that stays supple even at temperature extremes of -70 to 150°C. Remarkably durable, moisture, heat, and flame resistant. Provided with an elastic cinch for a no-slip fit.
历史批次信息供参考:
分析证书(COA)