SYNF901410
别名:
Synthware F901410, modified Schlenk reaction flask
1 of 10
Sigma-Aldrich
Z515981
Schlenk reaction and storage tube
Z166898
Aldrich® 气敏化学玻璃实验室套件
Z409235
Aldrich® Schlaker reaction tube
Z516082
Z515698
Schlenk Kjeldahl reaction and storage flask
Z516007
SYNF891410
Synthware™ reaction tube with glass stopcock
SYND271420
Synthware™ distillation distribution adapter
Z552011
Aldrich® vacuum adapter with SafetyBarb® hose connector
SYNS401420
Synthware™ pennyhead stopper, hollow
borosilicate glass flaskpear-shaped single-neck
joint: ST/NS14/20
pkg of 1 ea
Synthware F901410
10 mL
正在寻找类似产品? 访问 产品对比指南
样品处理
从最新的版本中选择一种:
抱歉,我们目前尚未线上提供该产品的COA。
如需帮助,请联系 客户支持
在文件库中查找您最近购买产品的文档。
访问文档库
SYNF561025
Synthware™ reaction tube with high vacuum valve
Z220264
Reaction flask with glass stopcock on side-arm
Z173061
Solvent storage flask with glass stopcock on side-arm
Z220590
Solid-transfer vessel with two side-arms and a gas inlet with glass stopcock
SYNP160001
Synthware™ cylindrical pressure vessel, heavy wall
Z648299
微量称重勺和小铲套装
JKP2003R
4arm-PEG10K
Z181064
Ace 压力管
11590
4,4'-偶氮双(4-氰基戊酸)