SYNF461250
别名:
Synthware F461250, round flask with side arm
1 of 10
SYND271420
Synthware™ distillation distribution adapter
Sigma-Aldrich
QBD10009
Biotin-dPEG®4-TFP ester
Z231215
Aldrich® modified sublimation apparatus
SYND682440
Synthware™ distillation distribution adapter with vacuum hose connection
SYNP120050C
Synthware™ peptide synthesis vessel with fritted disc and PTFE stopcock
GF22030445
聚丙烯
SYNA302440
Synthware™ vacuum or argon/nitrogen adapter with hose connection on top
Z173088
Solvent storage flask with glass stopcock on side-arm
MP8350205
MakroPor, silicon membrane
SYND161420
Synthware™ cow style distillation receiver
borosilicate glass flaskround bottom single-neck
joint: ST/NS14/20
pkg of 1 ea
Synthware F461250
250 mL
0-4 mm
正在寻找类似产品? 访问 产品对比指南
实验室容器与储存
实验室烧瓶
样品处理
历史批次信息供参考:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our 文件 section.