尺寸
20G
包裝
pkg of 20 x 5 ea (5 needles per sterile Tyvek peel pouch - 20 pouches per box)
長度 × 直徑
2.8 in. × 1.9 mm , ball
正在寻找类似产品? 访问 产品对比指南
一般說明
- Designed to enable the researcher to shape the needle to specific needs
- Needles come straight and are easily bent to any desired shape
- Made of type 304 stainless steel tubing
- Plastic Luer hubs
- Silicone tips for smoothest possible insertion
- Packaged 5 needles per sterile Tyvek peel pouch - 20 pouches per box
其他客户在看
Global Trade Item Number
货号 | GTIN |
---|---|
CAD9916-100EA | 4061834363613 |