Key Documents
CAD9915
Animal feeding needle, sterile, disposable, malleable 304 SS
size 15G, L × diam. 3 in. × 2.9 mm , ball
登录查看公司和协议定价
About This Item
分類程式碼代碼:
42142609
NACRES:
NB.03
尺寸
15G
包裝
pkg of 20 x 5 ea (5 needles per sterile Tyvek peel pouch - 20 pouches per box)
長度 × 直徑
3 in. × 2.9 mm , ball
正在寻找类似产品? 访问 产品对比指南
一般說明
  • Designed to enable the researcher to shape the needle to specific needs
  • Needles come straight and are easily bent to any desired shape
  • Made of type 304 stainless steel tubing
  • Plastic Luer hubs
  • Silicone tips for smoothest possible insertion
  • Packaged 5 needles per sterile Tyvek peel pouch - 20 pouches per box
从最新的版本中选择一种:
分析证书(COA)