包裝
pack of 5 ea
製造商/商標名
Cadence Science 6513
正在寻找类似产品? 访问 产品对比指南
一般說明
- Plated brass or 316 stainless steel
- Meets ISO 594 specifications
- Final assembly water tested at 45 PSI
Female Luer Lock (FLL) - contains a tabbed hub for secure fitting to a threaded male Luer lock.
Micro-Mate® Female Luer Lock (MMFLL) - designed and manufactured to reduce the stripping of threads which occurs when metal adapters are mated with plastic male Luer lock adapters. Uniquely designed to facilitate easy cleaning and maintenance.
Male Luer Lock (MLL) - male Luer slip with a threaded sheath for a secure fitting to a female Luer lock.
Male Luer (ML) - male Luer slip that fits with a female Luer.
法律資訊
Micro-Mate is a registered trademark of Cadence Science
Global Trade Item Number
货号 | GTIN |
---|---|
CAD6513-5EA | 4061832517827 |