主要文件
BR4630241
BRAND® Dispensette® S 有机定量瓶口分液器
volume 10 mL, with recirculation valve
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121500
NACRES:
NB.01
特點
DE-M marking
with recirculation valve
製造商/商標名
BRAND 4630241
容積
10 mL
接頭
(GL 28/S 28)
GL 32
GL 38
S 40
正在寻找类似产品? 访问 产品对比指南
一般說明
Dispensette S Organic is ideal for dispensing of organic solvents and acids
特點和優勢
  • Extremely easy filling and dispensing
  • Simple priming by optimized flow channels
  • Rapid and safe volume adjustment with internal scalloped track
其他說明
Fixed Volume- For standard applications. Simple calibration adjustment with a supplied tool.
法律資訊
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
历史批次信息供参考:
分析证书(COA)
访问文档库
实验方案
This article provides testing instructions for BRAND Dispensette® S bottle-top liquid dispenser according to the standard ISO DIS 8655.
This article provides testing instructions for BRAND Dispensette® S bottle-top liquid dispenser according to the standard ISO DIS 8655.