Merck
TEM-CF400CUUL
Sigma-Aldrich
Ultrathin Carbon Film Supported Copper Square Mesh
size 400 mesh, box of 50
Synonym(e):
CARBON SQUARE MESH - CU 400# UL 50/BX
About This Item
UNSPSC-Code:
41111761
NACRES:
MA.24
Verpackung
box of 50
Größe
400 mesh
Allgemeine Beschreibung
A 3-4 nm ultrathin film of pure carbon supported standard squre mesh grids. Copper grid, 400 mesh.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)