Documenti fondamentali
2879825-U
Supelco
Inlet Liner, Split/Splitless Type, Straight FocusLiner Design (wool packed)
pkg of 25 ea
Sinonimo/i:
GC accessory, glass wool, inlet liner
About This Item
Codice UNSPSC:
41115719
NACRES:
SB.54
Confezionamento
pkg of 25 ea
Lungh. × D.E. × D.I.
78.5 mm × 6.3 mm × 4.0 mm
Descrizione generale
Deactivated (silanized) glass inlet liner for capillary injection ports.
  • Fits Agilent (5890, 6890, and 7890)
  • Fits Finnigan (9001GCQ)
  • Fits Varian (CP-1177 Injector)
Note legali
FocusLiner is a trademark of SGE Analytical Science Pty Ltd.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)