Documenti fondamentali
22989
Supelco
Stainless Steel HPLC Fittings
ferrule, 1/8 in. tubing
Sinonimo/i:
Valco® FERRULE 1/8 PK 10
About This Item
Codice UNSPSC:
41115700
Materiali
303 stainless steel
Confezionamento
pkg of 10 ea
Produttore/marchio commerciale
VICI Valco® ZF2
Compatibilità
1/8 in. tubing
Prodotti consigliati
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Note legali
Valco is a registered trademark of Valco Instruments Co. Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 6
1 of 6
Swagelok® Union Swagelok® 200-6, stainless steel, 1/8 in., for connecting 1/8 to 1/8 in.
22041
Swagelok® Union
L-Asparagine certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
51363
L-Asparagine
PEEK Tubing L × O.D. × I.D. 10 ft × 1/8 in. × 0.062 in., natural
54995
PEEK Tubing
Tube Fitting, Cheminert® 1/4-28 male UNF for 1/8 in. O.D. tubing 3 mm (includes washer), (natural color)
58711
Tube Fitting, Cheminert®
Supelco® PEEK Fitting One-piece fingertight, pkg of 5 ea
Z227250
Supelco® PEEK Fitting