Documenti fondamentali
22146-U
Supelco
Gas Sampling Bulb
size 125 mL, glass stopcock
About This Item
Codice UNSPSC:
41104008
NACRES:
NB.24
Materiali
glass stopcock
tecniche
whole air sampling: suitable
Misura
125 mL
applicazioni
air monitoring
environmental
industrial hygiene
Descrizione generale
Use these glass bulbs to trap and transport a gas sample, then remove an aliquot (through the plug-type septum) for analysis. Oven annealed to resist damage during transportation and on-site use. Available with PTFE or high vacuum glass stopcocks. One half-hole cylindrical Thermogreen septum is included with each bulb.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)