Documenti fondamentali
Z757780
Sartorius filter pipette tips
volume range 2-120 μL, sterile, pack of 960 ea (10 racks of 96 tips)
Sinonimo/i:
filter pipette tips, pipetting tips, pipettor tips
About This Item
Codice UNSPSC:
41121601
NACRES:
NB.01
Intervallo di volume:
2-120 μL
Sterilità:
sterile
Caratteristiche:
barrier
filter
Confezionamento:
pack of 960 ea (10 racks of 96 tips)
Produttore/marchio commerciale:
Sartorius 790101F
Materiali
colorless
polypropylene (Virgin)
Sterilità
sterile
Caratteristiche
barrier
filter
Confezionamento
pack of 960 ea (10 racks of 96 tips)
Produttore/marchio commerciale
Sartorius 790101F
Intervallo di volume
2-120 μL
Tipo di binding
low binding surface
Descrizione generale
The Sartorius filter tips of the Biohitfamily are made of polypropylene and feature a polyethylene filter barrier to prevent aerosol and liquid contamination. This filter helps protect against the rist of cross-contamination and reduces maintenance requirements of the pipette. The filter tips are suited for molecular biology, microbiology, cell culture and radioactive work.
These tips have more space between the sample and the filter than conventional filter tips so you do not need to worry about the sample touching the filter whatever the pipetting technique or type of liquid.
Benefits:
  • Pipette foaming liquids like buffers and proteins
  • Pipette solvents
  • Reverse pipetting
  • Multiple dispensing functions of electronic pipettes
Altre note
Browse all pipette tips
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)