Documenti fondamentali
Z743553
Thermal transfer labels, Xylene resistant
L × W 1.69 in. × 0.625 in., roll of 1000
Sinonimo/i:
solvent resistant labels, temperature resistant labels
About This Item
Codice UNSPSC:
55121611
NACRES:
NB.35
Confezionamento
roll of 1000
Produttore/marchio commerciale
Diversified Biotech XYLN-2000
Lungh. × largh.
1.69 in. × 0.625 in.
Descrizione generale
Xylene resistant labels formatted for direct thermal printers. Resistant to most laboratory solvents and have short-term resistance to Acetone and xylene. When direct thermally printed, labels exhibit exceptional durability. Easily remove from surfaces without leaving any adhesive residue. Great for beakers, bottles, flasks, glass vials, tubes, tube racks and pipettes.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)