Key Documents
Z740269
Thermal transfer labels, Xylene resistant
pkg of 1000 ea (per roll)
Sinonimo/i:
solvent resistant labels, temperature resistant labels
About This Item
Codice UNSPSC:
55121611
NACRES:
NB.35
Materiali
polyester
white
Confezionamento
pkg of 1000 ea (per roll)
Produttore/marchio commerciale
Diversified Biotech XYLN-1000
Misura
0.875 in. × 0.875 in.
Descrizione generale
Temperature Resistant - From -40°C to 150°C
Barcoding - Print popular barcode formats for fast and easy sample identification
Printing Flexibility - As little as one label at a time, eliminating label waste and saving time and money
Superior - Unique solvent-resistant, acrylic-based adhesive provides excellent cold temperature performance
Tough - Resists degradation from scuffing, chemicals, moisture and wide temperature fluctuations
Specially Desigined - Coated for use with a resin ribbon that provides the best performance under extreme conditions
High Resolution - Print crisp, clear, professional quality labels