Z743536
Richter Anti-Vibration pads
L × W 8.5 in. × 11.0 in., pkg of 2
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.35
Prodotti consigliati
Confezionamento
pkg of 2
Produttore/marchio commerciale
Diversified Biotech VIBE-2000
Lungh. × largh.
8.5 in. × 11.0 in.
Descrizione generale
Inexpensive solution which dampens vibrations generated from lab equipment
Creates a stable surface to keep equipment in place
Fits vortexes, shakers, rockers and microscopes
Small, lightweight design
Stack pads for heavier units or position side-by-side forlarger equipment
Creates a stable surface to keep equipment in place
Fits vortexes, shakers, rockers and microscopes
Small, lightweight design
Stack pads for heavier units or position side-by-side forlarger equipment
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)