Documenti fondamentali
Z722413
IKA® ULTRA TURRAX® Tubes
BMT-G Ball-Mill tube with glass balls, 25/cs
Sinonimo/i:
tissue grinders, tissue homogenizers
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.22
Confezionamento
case of 25
pack of 25
Produttore/marchio commerciale
IKA 3703300
Descrizione generale
IKA® ULTRA-TURRAX® Disperser Tubes.
These hermetically sealed tubes prevent the spreading of aerosols or vapours created by mechanical stirring, dispersion or grinding infectious, toxic or odourous substances in open vessels. Disposable and single-use tubes, they avoid the possibility of cross-contamination of samples and remove the need for clean up of disperser tools, stirrer tools, grinding chambers or vessels.
Each tube has a bayonet fitting on the bottom, for easy, secure connection to the tube drive.
Note: Complete with glass balls for dry milling or cell disruption.
Attenzione
Stirring, dispersing or grinding may generate heat or cause other changes in the sample that might lead to build up of pressure inside the tube. Use appropriate personal protective equipment when operating and opening tubes.
Note legali
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
ULTRA-TURRAX is a registered trademark of IKA-Werke GmbH & Co. KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)