Documenti fondamentali
Z693707
Duran® filtering flask, with Keck® assembly set
capacity 500 mL
Sinonimo/i:
212044453
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.43
Materiali
borosilicate glass
flat bottom flask
Giunto
joint ST/NS neck34 mm I.D.
Sterilità
non-sterile
Confezionamento
pkg of 1 ea
pkg of 10 ea
Produttore/marchio commerciale
DURAN® 212044453
Disponibilità
not available in USA and Canada
Capacità
500 mL
Descrizione generale
The Duran filtering flask includes a Keck clip assembly and is idea for vacuum filtrations. The plastic hose connections meet safety and product regulations, and the flask has the Erlenmeyer shape.
Manufactured from Duran borosilicate glass type 3.3, the flasks have maximum chemical and thermal resistance and meet ISO 6556 standard.
Note legali
DURAN is a registered trademark of DWK Life Sciences
Keck is a registered trademark of Adcuram Lab Technologies GmbH
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)