Z646709
BRAND® Transferpette® S starter kits
Macro
About This Item
Codice UNSPSC:
41121500
NACRES:
NB.01
Prodotti consigliati
Produttore/marchio commerciale
BRAND
BrandTech 704792
Descrizione generale
Contains a set of 3 Transferpette S, Digital adjustable volume, 3 filled Tip-Box N and 3 wall/rack mounts.
Includes Transferpette S models D-1000, D-5000 & D-10000
Caratteristiche e vantaggi
- Efficient: one-hand operation
- Flexible: ergonomic design for all hand sizes
- Reliable: volume-change protection prevents accidental adjustments
Altre note
BR704792 (India)
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Transferpette is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)