Documenti fondamentali
Z377597
Sigma-Aldrich
Gel drying frames
Extra cellophane sheets, size 14 cm × 14 cm
Sinonimo/i:
crack prevention gel drying rack, gel drying frame
About This Item
Codice UNSPSC:
41105300
NACRES:
NB.22
Confezionamento
pk of 50 sheets
Produttore/marchio commerciale
Diversified Biotech AB-110762
Misura
14 cm × 14 cm
Descrizione generale
Dry gels quickly and easily between sheets of cellophane without heat or vacuum. Simply equilibrate gel in 20% ethanol − 10% glycerol to reduce swelling, then assemble cellophane sheets and gel in a sandwich on the drying frame, and leave to dry at room temperature. Most gels dry completely overnight.
Extra cellophane sheets
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)