Z367826
Culture plate racks
The Stacks 150
About This Item
Codice UNSPSC:
41122800
NACRES:
NB.22
Prodotti consigliati
Sterilità
non-sterile
Caratteristiche
Holds 40 x 150 mm plates
Produttore/marchio commerciale
IBI ACCRL0001
Categorie correlate
Descrizione generale
Designed for use on the Belly Dancer orbital shaker, especially for amplification of cDNA libraries, or any application requiring rocking of culture plates. Made of clear acrylic and Delrin®, withstand temperature to 60°C. Convenient handle cut in center.
Note legali
Delrin is a registered trademark of E. I. du Pont de Nemours and Company
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)