Z364517
Nalgene® media bottles, style 2019
capacity 500 mL
Sinonimo/i:
bottle nalgene, nalgene containers, sterile bottles
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.17
Prodotti consigliati
Materiali
high-density polyethylene cap
square bottle
transparent PETG
Giunto
Joints threaded neck (38-430mm)
Descrizione
Not autoclavable
Sterilità
sterile; γ-irradiated
Caratteristiche
graduations
with cap
Confezionamento
pack of 24 ea (2 xpack of 12)
Produttore/marchio commerciale
Nalgene® 2019-0500
Capacità
500 mL
Descrizione generale
- Nalgene® media bottles are made from rigid and sturdy polyethylene terephthalate copolyester, glycol-modified (PETG).
- Designed for storing buffers, cultures, and culture media for various laboratory applications.
- Features a leakproof closure for sample protection.
- Heat-shrink band around closure and neck facilitate tamper-evident seal.
- Ideal for refrigerated/frozen storage down to -40°C.
Media bottles (square), with white HDPE screw closure. Low gas permeability, non-pyrogenic. Heat-shrink band around closure and neck provides tamper-evident seal.
Caratteristiche e vantaggi
- Avoids depyrogenation, expensive washing, and autoclaving steps
- Sterile and non-pyrogenic
- Obtained in low particulate format
- Validated by extensive documentation
- Useful for storing and sampling active pharmaceutical ingredients and bulk intermediates
- Suitable for preparation and containment of buffers, cultures or extended storage of pH-sensitive liquids such as culture media.
Note legali
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Possiedi già questo prodotto?