Documenti fondamentali
Z768618
Nalgene® square graduated media bottle
without cap, PET, sterile, capacity 1000 mL
Sinonimo/i:
media storage bottle, square plastic bottle
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.17
Materiali
PET
square bottle
Giunto
Joints threaded neck (38-430)
Sterilità
sterile
Caratteristiche
graduations
without cap
Confezionamento
pack of 24 ea (2 x pack of 12)
Produttore/marchio commerciale
Nalgene® 342044-1000 (USA)
Nalgene® 842044-1000 (European)
Capacità
1000 mL
Descrizione generale
“Nalgene” Square PET media bottles offer sterility, lightweight construction, and shatter resistance. They are specifically designed for use in filling lines.
Caratteristiche e vantaggi
  • Has molded-in graduations and neck-ring.
  • Non-cytotoxic, non-hemolytic, and non-pyrogenic.
  • Exhibits exceptional gas barrier properties.
  • Gamma irradiated to 10^-6 to achieve sterility assurance level (SAL).
  • Composed of material free from ADC (animal derived component).
Altre note
Closures for these bottles are sold separately.
Note legali
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)