Key Documents
Z333972
BRAND® pipette fillers
FLIP, for pipettes up to 100 mL
About This Item
Codice UNSPSC:
41121500
NACRES:
NB.01
Materiali
natural rubber
Descrizione
2 valves
Caratteristiche
FLIP
for pipettes up to 100 mL
Produttore/marchio commerciale
BRAND 25400
Volume
100 mL
Descrizione generale
Simple pipetting aids for volumetric and graduated pipettes. Control of the functions by squeezing the appropriate valves between thumb and fore finger.
Altre note
BR25400 (India)
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 5
1 of 5
BRAND® dropping pipettes capacity 1.5 mL, with integrated bellows
Z331783
BRAND® dropping pipettes
Atomizer bulb useful for priming liquid-transfer lines
Z186724
Atomizer bulb
BRAND® pipette withdraw volume 5.5 mL (including bulb), graduations, 0.25 mL
BR747775
BRAND® pipette