Documenti fondamentali
WPB01041A
Whirl-Pak® Sampling Spoons
sterile
Sinonimo/i:
dispensing tool
About This Item
Codice UNSPSC:
24111500
NACRES:
NB.35
Materiali
polystyrene
Sterilità
sterile
Confezionamento
pkg of 100 ea
Produttore/marchio commerciale
Nasco Whirl-pak® B01041A
Lunghezza
~18 cm
Capacità
~15 mL
Descrizione generale
This sterilized spoon is ideal for use with Whirl-Pak® bags where sterile techniques are required for collection of samples.
Caratteristiche e vantaggi
  • Single-time use removes contamination risk
  • The easy-to-use spoon is made of sturdy, durable polystyrene
  • The procedure for contamination-free use is to open the bag, push one spoon up, pull it out, and reclose the bag
Note legali
Whirl-pak is a registered trademark of The Aristotle Corporation