BAH369300000
Sterileware™ sampling spatula
polystyrene, sterile
Sinonimo/i:
laboratory spoon
About This Item
Codice UNSPSC:
41122403
NACRES:
NB.35
Prodotti consigliati
Materiali
polystyrene
Sterilità
sterile
Confezionamento
pack of 100 ea
Produttore/marchio commerciale
Bel-Art H36930-0000
Lungh. × Largh. × Alt.
152 mm (6 in.) × 19 mm (3/4 in.) × 13 mm (1/2 in.)
Lungh. complessiva
9 in.
Categorie correlate
Descrizione generale
One-piece spatula is molded from rigid, white polystyrene and retains approximately 15cc of material. Shape allows easy transfer of sample to container. Use to sample directly from production line or containers including bags and drums.
- V-shaped spatula is perfect for sampling granulated or semi solid material
- Sterile and individually packaged
- One time use averts cross contamination
- Sharp point will pierce most packaging
Note legali
Sterileware is a trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)