TMO5921-0060
Nalgene® petri dish rack
for 60 mm petri dishes, polycarbonate, 8 positions, case of 2 ea
Sinonimo/i:
dish holder, plastic rack
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.13
Prodotti consigliati
Materiali
polycarbonate
Confezionamento
case of 2 ea
Produttore/marchio commerciale
Nalgene 5921-0060
Descrizione generale
Secure glass and plastic dishes for safe transport with Nalgene Petri and Bioassay Dish Racks.
Note legali
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)