Documenti fondamentali
TMO374500-BR
Nunc® Universal Tubes in 2D bar coded latch rack
polypropylene, capacity (1.8 mL), flat bottom, external thread, untreated, case of 10 × 24 ea
Sinonimo/i:
bar coded tube storage, traceable tube storage
About This Item
Codice UNSPSC:
41122800
NACRES:
NB.11
Materiali
flat bottom
polypropylene
Confezionamento
case of 10 × 24 ea
Produttore/marchio commerciale
Nunc 374500-BR
Capacità
(1.8 mL)
Categorie correlate
Descrizione generale
Experience the best features of the Thermo Scientific storage tube portfolio: universal cap, universal latch rack, and free-standing tube. This versatile 2.0mL internally-threaded tube is ideal both for a large storage system like a biobank, and in smaller scale for traditional cryostorage. **Now available in a barcoded rack to allow tracking of the rack in addition to individual tubes. Sample identity is ensured due to a permanently attached 2D barcode on each tube and permanent laser etching of the rack. All tubes and racks are supplied free from DNA, RNase, DNase, and endotoxins.
Note legali
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)