Key Documents
RA481WO-EP
Rotor for Eppendorf® 5804/5804R & 5810/5810R Centrifuges
A-4-81, for 5810/5810R, aerosol-tight, without buckets
About This Item
Codice UNSPSC:
41103909
Caratteristiche
to hold 4 x 500 mL rectangular buckets
to hold 4 x MTP/flex buckets
aerosol-tight
for 5810/5810R
medium to high-capacity swing-bucket rotor
without buckets
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Eppendorf® 022638611 (NA)
Eppendorf® 5810743001 (EU)
Parametri
4,000 max. RCF (3,220 xg)
tecniche
centrifugation: suitable
Compatibilità
for use with centrifuge models 5804/5804R/5810/5810R
Categorie correlate
Note legali
Eppendorf is a registered trademark of Eppendorf AG