Documenti fondamentali
OH30456389
Ohaus® Navigator® Portable Balance
Model NVT2201, weighing capacity 2200 g, resolution: 0.1 g, AC/DC input 230 V, universal plug set
About This Item
Codice UNSPSC:
41111502
NACRES:
NB.71
Materiali
metal weighing plate
Caratteristiche
CE compliant
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Ohaus® 30456389
Capacità di pesata
2200 g, resolution: 0.1 g
Largh. × Lungh. della navicella
17.4 cm × 23 cm
Descrizione generale
OHAUS raises the bar in price-performance ratio with the newly-designed Navigator series which is perfect for general weighing applications in laboratory, industrial and education settings. Ruggedly constructed and multifunctional, the Navigator′s a powerful balance that can handle a diverse range of weighing applications with features such as fast stabilization time, overload protection at four times its rated capacity, and easy operation.
Applicazioni
Weighing, Parts Counting, Percent Weighing, Checkweighing
Caratteristiche e vantaggi
Menu and calibration lock switch, mechanical and software overload/underload protection, stability indicator, low battery indicator, auto shut-off, user-selectable print options, user-selectable communication settings
Note legali
Navigator is a registered trademark of Ohaus Corp.
Ohaus is a registered trademark of Ohaus Corp.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)